Learn More
LGC GENOMICS LLC (Lucigen) Red Cell Lysis Solution, 1200 mLSelectively lyse red blood cells while leaving white blood cells intact. To be used with the MasterPure Kits.
Catalog No. NC2162068
Encompass
Save: Reduce costs by purchasing these important reagents in bulk quantitiesScale-up: Use these large volumes of reagents to perform large-scale MasterPure purifications
This item is not returnable. View return policy
Catalog No. NC2162068 Supplier LGC GENOMICS LLC Supplier No. MRC0912H
Save: Reduce costs by purchasing these important reagents in bulk quantitiesScale-up: Use these large volumes of reagents to perform large-scale MasterPure purifications