Learn More
Leica Biosystems RED COUNTERSTAIN
Catalog No. NC1940152
Encompass
Red Counterstain is designed to be used in conjunction with BOND Polymer Refine HRP Plex Detection kit (DS9914) and our chromogen reagents. This counterstain is a fast nuclear red stain which provides excellent contrast against a variety of chromogens, especially when used with the Blue Chromogen (DC9896) and Green Chromogen (DC9913).Red Counterstain is supplied in a volume of 15 mL.
Catalog No. NC1940152 Supplier Leica Biosystems Supplier No. AR9895
Red Counterstain is designed to be used in conjunction with BOND Polymer Refine HRP Plex Detection kit (DS9914) and our chromogen reagents. This counterstain is a fast nuclear red stain which provides excellent contrast against a variety of chromogens, especially when used with the Blue Chromogen (DC9896) and Green Chromogen (DC9913).Red Counterstain is supplied in a volume of 15 mL.