Learn More
Andwin Scientific Red Cryogenic labels 1.28" x 0.50", 1700 pack
SDP
Catalog No. NC9760007
Encompass_Preferred
Cryogenic labels formatted for laser printers. The 1.28" x 0.50" Cryo-Babies labels can withstand most organic solvents, caustic agents, temperatures from -196C to 100C. This product can still be utilized while under oven, autoclaving, or extreme freezer temperatures. The labels can also be used on several surfaces like plastics, glass, and metals without cracking, peeling, or degrading. This product is sold in the color of red.
Catalog No. NC9760007 Supplier Andwin Scientific Supplier No. LCRY1700R
Add to Cart
Add to Cart
Cryogenic labels formatted for laser printers. The 1.28" x 0.50" Cryo-Babies labels can withstand most organic solvents, caustic agents, temperatures from -196C to 100C. This product can still be utilized while under oven, autoclaving, or extreme freezer temperatures. The labels can also be used on several surfaces like plastics, glass, and metals without cracking, peeling, or degrading. This product is sold in the color of red.