Learn More
Key Scientific Products CryoCare, Organism Preservative System, Red Beads, 50 Vials per Box
SDP
Catalog No. NC9693417
Encompass_Preferred
CryoCare organism preservative system lowers the cost of stock cultures by stretching each expensive culture into 15 or more which meet NCCLS standards. Vial contains ceramic beads in a broth base. Includes 50 vials per box with red beads.
Catalog No. NC9693417 Supplier Key Scientific Products Supplier No. KS70RE50
CryoCare organism preservative system lowers the cost of stock cultures by stretching each expensive culture into 15 or more which meet NCCLS standards. Vial contains ceramic beads in a broth base. Includes 50 vials per box with red beads.