Learn More
Med Vet International Thermometer, Total Immersion, -20° to 150° C, 10/Box
Catalog No. 503046897
Encompass_Preferred
Non-toxic red-filled thermometers can be used in most laboratory applications. Anti-roll device included. Box of 10
Catalog No. 50-304-6897 Supplier Med Vet International Supplier No. 973111BX
Non-toxic red-filled thermometers can be used in most laboratory applications. Anti-roll device included. Box of 10