Learn More
Cospheric LLC FMR - Red Fluorescent Microspheres 1.3g/cc 1-5um - 500mg
SDP
Catalog No. NC1814397
Encompass
Bright red fluorescent spherical polymer particles in diameters between 1-5microns are supplied in dry powder form and are color-stable in solution. Small fluorescent microspheres have numerous applications in biotechnology, medical research, medical devices, diagnostics, process troubleshooting and other fields of study.
Catalog No. NC1814397 Supplier Cospheric LLC Supplier No. FMR1.3 15UM500MG
Bright red fluorescent spherical polymer particles in diameters between 1-5microns are supplied in dry powder form and are color-stable in solution. Small fluorescent microspheres have numerous applications in biotechnology, medical research, medical devices, diagnostics, process troubleshooting and other fields of study.