Learn More
Apexbio Technology LLC Red Fluorometric Lipid Droplet Assay Kit 500T
SDP
Catalog No. 5000097189
Encompass_Preferred
Red Fluorometric Lipid Droplet Assay Kit is a fluorescent detection kit designed for the visualization and quantification of intracellular lipid droplets It is intended to probe and monitor lipid metabolism by specifically labeling intracellular neutral lipid accumulations This kit exerts its function primarily through the use of Nile Red a cell-permeable lipophilic dye that selectively accumulates within neutral lipid environments resulting in red fluorescence upon excitation (552 nm emission 636 nm) No key metric (IC50/EC50) or specific cell line information is available Based on these properties Red Fluorometric Lipid Droplet Assay Kit enables research applications in cellular lipid metabolism autophagy cell differentiation and senescence
Catalog No. 50-000-97189 Supplier Apexbio Technology LLC Supplier No. K2169500
Red Fluorometric Lipid Droplet Assay Kit is a fluorescent detection kit designed for the visualization and quantification of intracellular lipid droplets It is intended to probe and monitor lipid metabolism by specifically labeling intracellular neutral lipid accumulations This kit exerts its function primarily through the use of Nile Red a cell-permeable lipophilic dye that selectively accumulates within neutral lipid environments resulting in red fluorescence upon excitation (552 nm emission 636 nm) No key metric (IC50/EC50) or specific cell line information is available Based on these properties Red Fluorometric Lipid Droplet Assay Kit enables research applications in cellular lipid metabolism autophagy cell differentiation and senescence