Learn More
Red-Four (TM) Buffer Solution, pH 4.01 +/- 0.01 at 25°C, Color-coded RED, Reference Standard, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Red-Four (TM) Buffer Solution, pH 4.01 ± 0.01 at 25°C, Color-coded RED, Reference Standard
Specifications
Specifications
| CAS | 877-24-7, 915-67-3, 7732-18-5 |
| CAS Max % | 0.01, 0.98 |
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 4.01 ±0.01 |
| Physical Form | Aqueous Solution |
| Quantity | 500 mL |
| Packaging | Poly Bottle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.