Learn More
Research Products International Corp Red-GAL [6-Chloro-3-indolyl-B-D-galactopyranoside], 1 Gram
SDP
Catalog No. 50488592
Encompass_Preferred
An alternative chromogenic substrate for β-D-galactosidase. Red-GAL is used in conjuction with IPTG for detection of β-galactosidase activity in bacterial colonies in a colorimetric assay to detect recombinants from non-recombinants.
Catalog No. 50-488-592 Supplier Research Products International Corp Supplier No. C400401.0
An alternative chromogenic substrate for β-D-galactosidase. Red-GAL is used in conjuction with IPTG for detection of β-galactosidase activity in bacterial colonies in a colorimetric assay to detect recombinants from non-recombinants.