Learn More
Alkali Scientific Red GAL [6 Chloro 3 indolyl β D galactopyranoside], 100 Mg
SDP
Catalog No. 5000661703
Encompass_Preferred
Red GAL (6-Chloro-3-Indolyl β-D-Galactopyranoside) is a chromogenic substrate used for the detection of β-galactosidase activity in biochemical assays. This reagent produces a red color when hydrolyzed by β-galactosidase, making it ideal for monitoring gene expression in genetic studies and reporter assays. The 100mg quantity is perfect for smaller-scale experiments or routine use in laboratories studying enzyme activity and gene function. Red GAL is commonly used in bacterial screening, enzymatic assays, and molecular biology applications, providing a clear, colorimetric result for precise analysis of β-galactosidase activity in various samples.
Catalog No. 50-006-61703 Supplier Alkali Scientific Supplier No. RED001CP
Red GAL (6-Chloro-3-Indolyl β-D-Galactopyranoside) is a chromogenic substrate used for the detection of β-galactosidase activity in biochemical assays. This reagent produces a red color when hydrolyzed by β-galactosidase, making it ideal for monitoring gene expression in genetic studies and reporter assays. The 100mg quantity is perfect for smaller-scale experiments or routine use in laboratories studying enzyme activity and gene function. Red GAL is commonly used in bacterial screening, enzymatic assays, and molecular biology applications, providing a clear, colorimetric result for precise analysis of β-galactosidase activity in various samples.