Learn More
Braintree Scientific Red Gas Anesthetizing Box, 10"" x 4"" x 4""
SDP
Catalog No. 501954155
Encompass
Red Gas Anesthetizing Box with perforated floor for use with rodents. The .25" thick plastic anesthetizing box allows for constant observation while anesthetizing. Helps protect against accidental overdosing. For use with rats and mice. Easy to move machine tooled sliding top. Made of durable plastic, this box should last for years. Not for use with liquid organic solvents. Advantages of the red tube vs. the opaque tube Laboratory rodents are largely color blind, so a redbox simply looks like a comforting shelter. The Redbox is still transparent, allowing the investigator to observe the animal inside. Improved comfort for rats and mice during acute experimental procedures without sacrificing safety or welfare. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4155 Supplier Braintree Scientific Supplier No. ABRED1
 Restricted Item
Add to Cart
Add to Cart
Red Gas Anesthetizing Box with perforated floor for use with rodents. The .25" thick plastic anesthetizing box allows for constant observation while anesthetizing. Helps protect against accidental overdosing. For use with rats and mice. Easy to move machine tooled sliding top. Made of durable plastic, this box should last for years. Not for use with liquid organic solvents. Advantages of the red tube vs. the opaque tube Laboratory rodents are largely color blind, so a redbox simply looks like a comforting shelter. The Redbox is still transparent, allowing the investigator to observe the animal inside. Improved comfort for rats and mice during acute experimental procedures without sacrificing safety or welfare. **Sold for research purposes only and not for human use**