Learn More
Med Vet International Red Heparizined Microhematocrit Capillary Tubes, 75mm, J0543, 100/Tube, 10 Tubes/Box
Catalog No. 5000582332
Encompass_Preferred
These capillary tubes are designed for both safe blood collection as well as an accurate microhematocrit determination. Precision molded, they provide excellent sample retention during centrifugation. Tubes fill rapidly without affecting specimen integrity. Tube 1.15mm i.d. x 1.55mm o.d. 75mm length. Packaged in a protective snap cap vial. 100 tubes/vial/10 visls/cs
Catalog No. 50-005-82332 Supplier Med Vet International Supplier No. JOR543
Add to Cart
Add to Cart
These capillary tubes are designed for both safe blood collection as well as an accurate microhematocrit determination. Precision molded, they provide excellent sample retention during centrifugation. Tubes fill rapidly without affecting specimen integrity. Tube 1.15mm i.d. x 1.55mm o.d. 75mm length. Packaged in a protective snap cap vial. 100 tubes/vial/10 visls/cs