Learn More
Enzo Life Sciences Red hydrogen peroxide assay kit (1Kit)
SDP
Supplier:  Enzo Life Sciences ENZ51004
Encompass_Preferred
Red Hydrogen Peroxide Assay Kit provides a simple absorbance-based or fluorometric assay to detect and quantify hydrogen peroxide or peroxidase activity in biochemical assays, cell extracts and in live cells. It can also be used to detect a variety of oxidase activities through enzyme-coupled reactions. The kit is an optimized ‘mix and read’ assay that is suitable for both 96- and 348-well formats and is compatible with HTS liquid handling. Applications: Fluorescence microscopy, HTS .
Catalog No. 50-201-2577