Learn More
World Precision Instrument Red Insulated Mini Banana Plug
SDP
Catalog No. 502521495
Encompass
Common connector for WPI amplifiers
Banana plugs are often used for making connections with WPI amplifiers. This is a mini, insulated, male banana plug connector. The wire is held in place with an internal set screw.  The jack and plug are gold-plated to resist corrosion and provide optimal electrical connectivity.
Catalog No. 50-252-1495 Supplier World Precision Instrument Supplier No. 2034
Common connector for WPI amplifiers
Banana plugs are often used for making connections with WPI amplifiers. This is a mini, insulated, male banana plug connector. The wire is held in place with an internal set screw.  The jack and plug are gold-plated to resist corrosion and provide optimal electrical connectivity.