Learn More
MSC Red Kap Slash Pocket Jacket, Lined, Insulated, Twill, 1 ea
Catalog No. 5000795239
Encompass
This lined, insulated jacket is designed to keep you warm when working outdoors, offering protection against wind and rain. It features two inset slash pockets and a utility pocket on the left sleeve for convenient storage of necessities. The jacket is adjustable and practical, ensuring you can get the job done regardless of the weather.
  • Plenty of pockets, including two lower inset slash pockets and a utility pocket on left sleeve
  • Wrinkle resistant finish for a neat appearance
  • Black quilted lining provides added warmth
  • Traditional waist-length design
  • Adjustable waistband tabs for a customizable fit
  • Industrial laundry friendly for rigorous cleaning
  • Durable shell made of 7.25 oz. twill, 65% polyester and 35% cotton
  • Solid brass zipper for reliable closure
  • Water repellent fabric
Catalog No. 50-007-95239 Supplier MSC Supplier No. 76002161
This lined, insulated jacket is designed to keep you warm when working outdoors, offering protection against wind and rain. It features two inset slash pockets and a utility pocket on the left sleeve for convenient storage of necessities. The jacket is adjustable and practical, ensuring you can get the job done regardless of the weather.
  • Plenty of pockets, including two lower inset slash pockets and a utility pocket on left sleeve
  • Wrinkle resistant finish for a neat appearance
  • Black quilted lining provides added warmth
  • Traditional waist-length design
  • Adjustable waistband tabs for a customizable fit
  • Industrial laundry friendly for rigorous cleaning
  • Durable shell made of 7.25 oz. twill, 65% polyester and 35% cotton
  • Solid brass zipper for reliable closure
  • Water repellent fabric