Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Red light-emitting spiro-copolymer

Catalog No. 900446250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-044-6250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-044-6250MG Supplier Sigma Aldrich Supplier No. 900446250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Features:Soluble spiro-copolymerLong lifetime with further big potential to improveSaturated red color coordinatorsLow operation voltageVery good viscosity behaviour in many organic solventsTypical Performance measured in standard device structure (ITO/PEDOT: PSS 20 nm/SPR-001 80 nm/Ba 6 nm/Ag 150 nm):Max. Efficiency: 1.90 Cd/AV_on: 2.00 VV @ 100 cd/m2: 3.3 VV @ 1000 cd/m2: 5.1 VMax. Power Eff.: 1.80 lm/WExternal Quantum Eff.: 2.44 %CIE1931 @ 100 cd/m2: (0.67, 0.33)DC Lifetime @ 100 cd/m2 @ RT: >20,000Product of EMD Performance Materials Corp., an Affiliate of Merck KGaA, Darmstadt, Germany

Specifications

CAS 1365250-36-7
Quantity 250 mg
Synonym Livilux(TM) SPR-001
Molecular Weight (g/mol) 120000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.