Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Red light-emitting spiro-copolymer

Catalog No. 900444250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-044-4250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-044-4250MG Supplier Sigma Aldrich Supplier No. 900444250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Features:Soluble redspiro-copolymer.Long lifetime, with further big potential to improve.Saturated red color coordinators.Low operation voltage.Very good viscosity behaviour in many organic solvents.Typical performance measured in standard device structure (ITO/PEDOT:PSS 20 nm/SPR-001 80 nm/Ba 6 nm/Ag 150 nm):Max. efficiency: 1.90 Cd/A.V_on: 2.00 V.V at 100 cd/m2: 3.3 V.V at 1000 cd/m2: 5.1 V.Max. power eff.: 1.80 lm/W.External quantum eff.: 2.44%.CIE1931 at 100 cd/m2: (0.67, 0.33).DC lifetime at 100 cd/m2 at RT: >20,000.Product of EMD Performance Materials Corp., an Affiliate of Merck KGaA, Darmstadt, Germany

Specifications

CAS 1365250-36-7
Quantity 250 mg
Synonym Livilux(TM) SPR-001
Molecular Weight (g/mol) 50000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.