Learn More
Master Lock RED LOCK KEYED-ALIKE
Catalog No. NC2257845
Encompass_Preferred
American Lock Rekeyable Padlocks, Clearance: 0.75 in., 1.9cm, KeyType: Different, Aluminum, Height Shackle: 1 in., 2.54cm, Red,Certifications/Compliance: OSHA 1910.147, Diameter: 0.25 in., 0.6cmVendor Catalog # A1105KARED
Catalog No. NC2257845 Supplier Master Lock Supplier No. A1105KARED
Add to Cart
Add to Cart
American Lock Rekeyable Padlocks, Clearance: 0.75 in., 1.9cm, KeyType: Different, Aluminum, Height Shackle: 1 in., 2.54cm, Red,Certifications/Compliance: OSHA 1910.147, Diameter: 0.25 in., 0.6cmVendor Catalog # A1105KARED