Learn More
Enzo Life Sciences CYTO-ID Red long-term cell tracer kit (1Kit)
SDP
Catalog No. 502012600
Encompass_Preferred
The CYTO-ID Red Long-Term Cell Tracer Kit uses proprietary noncovalent cell labeling technology to stably incorporate a red fluorescent dye containing hydrophobic aliphatic chains into the cell membrane’s lipid bilayer. The dye may be loaded into cells by following the included protocol. The labeling buffer is isotonic for mammalian cells and contains no detergents or organic solvents. The appearance of labeled cells may vary depending upon the cell type from uniformly bright to punctuate. This difference is thought to relate to the extent of membrane internalization occurring after cell labeling. The CYTO-ID Red Tracer Dye fluorescence is independent of pH within normally encountered physiologic ranges and fluorescence intensity per cell is typically unaffected by the ultimate pattern of dye distribution. Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection .
Catalog No. 50-201-2600 Supplier Enzo Life Sciences Supplier No. ENZ51037K025
The CYTO-ID Red Long-Term Cell Tracer Kit uses proprietary noncovalent cell labeling technology to stably incorporate a red fluorescent dye containing hydrophobic aliphatic chains into the cell membrane’s lipid bilayer. The dye may be loaded into cells by following the included protocol. The labeling buffer is isotonic for mammalian cells and contains no detergents or organic solvents. The appearance of labeled cells may vary depending upon the cell type from uniformly bright to punctuate. This difference is thought to relate to the extent of membrane internalization occurring after cell labeling. The CYTO-ID Red Tracer Dye fluorescence is independent of pH within normally encountered physiologic ranges and fluorescence intensity per cell is typically unaffected by the ultimate pattern of dye distribution. Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection .