Learn More
AVANTOR MASTERFLEX (VWR LLC) RED.NIPPLE 1/2FTFTX1/4FTFTMPT
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4062351
Encompass
Masterflex Fitting, Polypropylene, Straight, Thread Reducer, 1/4IN NPT(M) 1/2IN NPT (M); 10/PK, Avantor
Catalog No. 50-274-2283