Learn More
AVANTOR MASTERFLEX (VWR LLC) RED.NIPPLE 1/2IN X1/4IN MPT 6
Catalog No. 502742669
Encompass
Masterflex Fitting, Nylon, Straight, Threaded Reducer, 1/4IN NPT(M) 1/2IN NPT (M); 6/PK, Avantor
Catalog No. 50-274-2669 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3062350
Masterflex Fitting, Nylon, Straight, Threaded Reducer, 1/4IN NPT(M) 1/2IN NPT (M); 6/PK, Avantor