Learn More
Montana Molecular Red Nucleus-targeted cADDis cAMP Assay - 10mL
SDP
Catalog No. NC2600266
Encompass_Preferred
Measure real-time cAMP signaling kinetics in the nucleus cilia with this live cell fluorescent assay. Add NLS-tagged cADDis to your cells and measure robust signals the next day on a fluorescence imaging system. Increasing red fluorescence indicates cAMP increase. This assay is genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in many primary or iPSC-derived cells. The kit includes the cAMP sensor BacMam, 500mM sodium butyrate in H2O, and a positive control receptor and agonist (B2AR BacMam and 10mM isoproterenol in HCl). Other Montana Molecular tools including BacMam-packaged GPCRs, green fluorescent membrane-targeted or cytosolic cAMP sensors, or green fluorescent sensors for DAG or Calcium can easily be co-expressed with this assay.
Catalog No. NC2600266 Supplier Montana Molecular Supplier No. U0221R010
Measure real-time cAMP signaling kinetics in the nucleus cilia with this live cell fluorescent assay. Add NLS-tagged cADDis to your cells and measure robust signals the next day on a fluorescence imaging system. Increasing red fluorescence indicates cAMP increase. This assay is genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in many primary or iPSC-derived cells. The kit includes the cAMP sensor BacMam, 500mM sodium butyrate in H2O, and a positive control receptor and agonist (B2AR BacMam and 10mM isoproterenol in HCl). Other Montana Molecular tools including BacMam-packaged GPCRs, green fluorescent membrane-targeted or cytosolic cAMP sensors, or green fluorescent sensors for DAG or Calcium can easily be co-expressed with this assay.