Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Red pigment-concentrating hormone | 37933-92-9 | 96.86% | 930.02 | 1 MG
SDP

Catalog No. 5000397403
Encompass_Preferred

Red pigment-concentrating hormone is a chromatophorotropic hormone and is synthesized in the eyestalk. It may play a role as a downstream hormone of 5-HT.

  • Appearance: Solid
  • Color: White to off-white
  • Sequence: {Glp}-Leu-Asn-Phe-Ser-Pro-Gly-Trp-NH2
  • Sequence shortening: {Glp}-LNFSPGW-NH2
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Sealed storage, away from moisture. Powder: -80°C for 2 years, -20°C for 1 year. In solvent: -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture)
  • Solubility (in vitro): DMSO: 100 mg/mL (107.52 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)

Catalog No. 50-003-97403 Supplier Medchemexpress LLC Supplier No. HYP38241MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Red pigment-concentrating hormone is a chromatophorotropic hormone and is synthesized in the eyestalk. It may play a role as a downstream hormone of 5-HT.

  • Appearance: Solid
  • Color: White to off-white
  • Sequence: {Glp}-Leu-Asn-Phe-Ser-Pro-Gly-Trp-NH2
  • Sequence shortening: {Glp}-LNFSPGW-NH2
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Sealed storage, away from moisture. Powder: -80°C for 2 years, -20°C for 1 year. In solvent: -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture)
  • Solubility (in vitro): DMSO: 100 mg/mL (107.52 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.