Learn More
Research Products International Corp Red Polyethylene Liners Printed with Biohazard Symbol, 2 mil Thick, 15 x 9 x 32 Inches, 25 per Package
SDP
Catalog No. 501978392
Encompass
Bright red, autoclavable bags feature one long flap for easy opening. Printed with the biohazard symbol and sterilization indicator patch, they are made of High Molecular Weight, High-Density (HMHD) polyethylene. Ensure safe handling and disposal of biohazardous waste. Fits a variety of containers and holders.
Catalog No. 50-197-8392 Supplier Research Products International Corp Supplier No. 17722
Bright red, autoclavable bags feature one long flap for easy opening. Printed with the biohazard symbol and sterilization indicator patch, they are made of High Molecular Weight, High-Density (HMHD) polyethylene. Ensure safe handling and disposal of biohazardous waste. Fits a variety of containers and holders.