Learn More
Bellco Glass, Inc. Red Pouring Ring,45mm
SDP
Catalog No. 501945822
Encompass_Preferred
Use with Hi-Temperature Red Cap. Pouring ring provides drip free pouring an protects thread from chipping. They are autoclavable to 180C. Packed 10 per case. MADE IN THE USA
Catalog No. 50-194-5822 Supplier Bellco Glass, Inc. Supplier No. 563791045
Use with Hi-Temperature Red Cap. Pouring ring provides drip free pouring an protects thread from chipping. They are autoclavable to 180C. Packed 10 per case. MADE IN THE USA