Learn More
Andwin Scientific RED PRESS/VACUUM RUBBER 50 FT
SDP
Catalog No. NC9451993
Encompass_Preferred
Red Pressure/Vacuum Rubber Tubing, Natural rubber tubing with extra-heavy wall for vacuum work. High resilience, good tensile and tear strength. Durometer hardness: Shore A, 50. Tensile strength: 2500 to 3500 Psi. Coil length: 15.2m
Catalog No. NC9451993 Supplier Andwin Scientific Supplier No. 62995062
Red Pressure/Vacuum Rubber Tubing, Natural rubber tubing with extra-heavy wall for vacuum work. High resilience, good tensile and tear strength. Durometer hardness: Shore A, 50. Tensile strength: 2500 to 3500 Psi. Coil length: 15.2m