Learn More
Perkin Elmer US LLC Red PTFE (Hydrophilic) Syringe Filters, 0.22 µm, 25 mm (100/pack)
Catalog No. 502113446
Encompass_Preferred
Hydrophilic PTFE syringe filters have broad chemical compatibility and high pH resistance and are optically clear when wet with water. They are versatile filters for use with aqueous and aggressive organic solvent-based solutions and are especially ideal for HPLC sample preparation. The pure polypropylene housings are color-coded and available in smooth rim or gear rim formats. All our syringe filters are HPLC certified. Housing Material: Polypropylene Effective Filtration Area (cm2): 4. 08 Pore Size (µm): 0. 22 µm Holdup Volume (µl): Sample Volume (mL): Inlet Connection: Female luer lock Outlet Connection: Male luer slip Maximum Operating Temperature: 50 °C Maximum Operating Pressure: 87 psi (6. 0 bar) at 20 °C Typical Applications: HPLC sample preparation Routine QA/QC analysis Content uniformity Removal of protein precipitates Dissolution testing Biofuel analysis
Catalog No. 50-211-3446 Supplier Perkin Elmer US LLC Supplier No. 02542926
Add to Cart
Add to Cart
Hydrophilic PTFE syringe filters have broad chemical compatibility and high pH resistance and are optically clear when wet with water. They are versatile filters for use with aqueous and aggressive organic solvent-based solutions and are especially ideal for HPLC sample preparation. The pure polypropylene housings are color-coded and available in smooth rim or gear rim formats. All our syringe filters are HPLC certified. Housing Material: Polypropylene Effective Filtration Area (cm2): 4. 08 Pore Size (µm): 0. 22 µm Holdup Volume (µl): Sample Volume (mL): Inlet Connection: Female luer lock Outlet Connection: Male luer slip Maximum Operating Temperature: 50 °C Maximum Operating Pressure: 87 psi (6. 0 bar) at 20 °C Typical Applications: HPLC sample preparation Routine QA/QC analysis Content uniformity Removal of protein precipitates Dissolution testing Biofuel analysis