Learn More
Perkin Elmer US LLC Red PTFE (Hydrophilic) Syringe Filters, 0.45 µm, 25 mm (100/pack)
Catalog No. 502113442
Encompass_Preferred
All our syringe filters are HPLC certified. Housing Material: Polypropylene Effective Filtration Area (cm2): 4.08 Pore Size (µm): 0.45 µm Holdup Volume (µl): <100 Sample Volume (mL): <100 Inlet Connection: Female luer lock Outlet Connection: Male luer slip Maximum Operating Temperature: 50 °C Maximum Operating Pressure: 87 psi (6.0 bar) at 20 °C Typical Applications: HPLC sample preparation Routine QA/QC analysis Content uniformity Removal of protein precipitates Dissolution testing Biofuel analysis
Catalog No. 50-211-3442 Supplier Perkin Elmer US LLC Supplier No. 02542927
Add to Cart
Add to Cart
All our syringe filters are HPLC certified. Housing Material: Polypropylene Effective Filtration Area (cm2): 4.08 Pore Size (µm): 0.45 µm Holdup Volume (µl): <100 Sample Volume (mL): <100 Inlet Connection: Female luer lock Outlet Connection: Male luer slip Maximum Operating Temperature: 50 °C Maximum Operating Pressure: 87 psi (6.0 bar) at 20 °C Typical Applications: HPLC sample preparation Routine QA/QC analysis Content uniformity Removal of protein precipitates Dissolution testing Biofuel analysis