Learn More
Perkin Elmer US LLC Red PTFE (Hydrophobic) Syringe Filters, 0.45 µm, 25 mm (100/pack)
Supplier:  Perkin Elmer US LLC 02542912
Encompass_Preferred
Hydrophobic PTFE syringe filters have broad chemical compatibility and high pH resistance. These are versatile filters for use with aggressive organic solvent-based solutions and are especially ideal for HPLC sample preparation. The pure polypropylene housings are color-coded and available in smooth rim or gear rim formats. All our syringe filters are HPLC certified. Housing Material: Polypropylene Effective Filtration Area (cm2): 4.08 Pore Size (µm):0.45 µm Holdup Volume (µl): <100 Sample Volume (mL): <100 Inlet Connection: Female luer lock Outlet Connection: Male luer slip Maximum Operating Temperature: 50 °C Maximum Operating Pressure: 87 psi (6.0 bar) at 20 °C Typical Applications: HPLC sample preparation Routine QA/QC analysis Content uniformity Removal of protein precipitates Dissolution testing Biofuel analysis
Catalog No. 50-211-3436