Learn More
Bellco Glass, Inc. Red PTFE/Silicone Septa,10mm
SDP
Catalog No. 501945396
Encompass_Preferred
Manufactured from red ptfe silicone. Ideal for screw thread vials. MADE IN THE USA. Autoclavable Size: 10mm Septa thickness: 0.060"
Catalog No. 50-194-5396 Supplier Bellco Glass, Inc. Supplier No. 207001600
Manufactured from red ptfe silicone. Ideal for screw thread vials. MADE IN THE USA. Autoclavable Size: 10mm Septa thickness: 0.060"