Learn More
Bellco Glass, Inc. Red PTFE/Silicone Septa,8mm
SDP
Catalog No. 501945390
Encompass_Preferred
Manufactured from red ptfe silicone. Ideal for screw thread vials. MADE IN THE USA. Autoclavable Size: 8mm Septa thickness: 0.065"
Catalog No. 50-194-5390 Supplier Bellco Glass, Inc. Supplier No. 207001000
Manufactured from red ptfe silicone. Ideal for screw thread vials. MADE IN THE USA. Autoclavable Size: 8mm Septa thickness: 0.065"