Learn More
Bellco Glass, Inc. Red PTFE/Slicone Septa,13mm
SDP
Catalog No. 501945392
Encompass_Preferred
Manufactured from red ptfe silicone. Ideal for screw thread vials. MADE IN THE USA. Autoclavable Size: 13mm Septa thickness: 0.075"
Catalog No. 50-194-5392 Supplier Bellco Glass, Inc. Supplier No. 207001200
Manufactured from red ptfe silicone. Ideal for screw thread vials. MADE IN THE USA. Autoclavable Size: 13mm Septa thickness: 0.075"