Learn More
MSC RED RAM WING THMB GLV
Catalog No. 5000767399
Encompass
These MCR Safety welding gloves are designed for comfort and protection, featuring a size large fit and constructed from a durable blend of leather and synthetic leather.
  • Welding glove
  • Numeric size 9
  • Size large
  • Primary material: synthetic leather and leather
  • Unlined for enhanced dexterity
  • Supplied as a pair
  • Constructed with pigskin leather
  • Clute glove back pattern
  • Country of origin: China
Catalog No. 50-007-67399 Supplier MSC Supplier No. 03570991
These MCR Safety welding gloves are designed for comfort and protection, featuring a size large fit and constructed from a durable blend of leather and synthetic leather.
  • Welding glove
  • Numeric size 9
  • Size large
  • Primary material: synthetic leather and leather
  • Unlined for enhanced dexterity
  • Supplied as a pair
  • Constructed with pigskin leather
  • Clute glove back pattern
  • Country of origin: China