Learn More
Andwin Scientific RED RUBBER BULB
SDP
Catalog No. NC9590603
Encompass_Preferred
The red rubber bulb is compatible for use with disposable Pasteur pipets. This product has a volume capacity of 2 mL, a bore size of 6 mm, a diameter of 13 mm, and a length of 32 mm.
Catalog No. NC9590603 Supplier Andwin Scientific Supplier No. 56310240
The red rubber bulb is compatible for use with disposable Pasteur pipets. This product has a volume capacity of 2 mL, a bore size of 6 mm, a diameter of 13 mm, and a length of 32 mm.