Learn More
Med Vet International Red Rubber Cuffed Endotracheal Tube 3.0mm, J0148B
Catalog No. 5000583673
Encompass_Preferred
Available in .5mm increments from 2.5 to 16.0mm. Murphy eye. Red rubber. Hard-to-find extra-large size now available. (adaptor not included except where specified)
Catalog No. 50-005-83673 Supplier Med Vet International Supplier No. JOR148B
Available in .5mm increments from 2.5 to 16.0mm. Murphy eye. Red rubber. Hard-to-find extra-large size now available. (adaptor not included except where specified)