Learn More
Med Vet International Red Rubber Cuffed Endotracheal Tube 5.5mm, J0148G
Catalog No. 5000583361
Encompass_Preferred
Available in .5mm increments from 2.5 to 16.0mm. Murphy eye. Red rubber. Hard-to-find extra-large size now available. (adaptor not included except where specified)
Catalog No. 50-005-83361 Supplier Med Vet International Supplier No. JOR148G
Available in .5mm increments from 2.5 to 16.0mm. Murphy eye. Red rubber. Hard-to-find extra-large size now available. (adaptor not included except where specified)