Learn More
Electron Microscopy Sciences Red Sable Brush Set of 3 Sizes (#2/0; #1 And #2)
SDP
Catalog No. 5024803
Encompass_Preferred
Extremely soft and pliable. Perfect for the manipulation of previously fixed, dehydrated specimens into embedding capsules or molds.
Catalog No. 50-248-03 Supplier Electron Microscopy Sciences Supplier No. 66105
Extremely soft and pliable. Perfect for the manipulation of previously fixed, dehydrated specimens into embedding capsules or molds.