Learn More
Electron Microscopy Sciences Brush, Red Sable, Size #1, 1 ea
SDP
Catalog No. 5024801
Encompass_Preferred
Extremely soft and pliable. Perfect for the manipulation of previously fixed, dehydrated specimens into embedding capsules or molds.
Catalog No. 50-248-01 Supplier Electron Microscopy Sciences Supplier No. 6610001
Extremely soft and pliable. Perfect for the manipulation of previously fixed, dehydrated specimens into embedding capsules or molds.