Learn More
Electron Microscopy Sciences Red Sable Brush Size # 2
SDP
Catalog No. 5024802
Encompass_Preferred
Extremely soft and pliable. Perfect for the manipulation of previously fixed, dehydrated specimens into embedding capsules or molds.
Catalog No. 50-248-02 Supplier Electron Microscopy Sciences Supplier No. 6610002
Extremely soft and pliable. Perfect for the manipulation of previously fixed, dehydrated specimens into embedding capsules or molds.