Learn More
Ted Pella Inc RED SABLE BRUSH SZ 3 3/PK
SDP
Catalog No. NC9266481
Encompass
Sable Brushes, Width at Ferrule 2mm, Length at Ferrule 13mm, no.3, Red, Sable hairs are springy and relatively fine, Enameled wood handles and seamless Nickel Ferrules
Catalog No. NC9266481 Supplier Ted Pella Inc Supplier No. 11816
Sable Brushes, Width at Ferrule 2mm, Length at Ferrule 13mm, no.3, Red, Sable hairs are springy and relatively fine, Enameled wood handles and seamless Nickel Ferrules