Learn More
Bioworld RED SEPAROPORE 4B-CL
Catalog No. 50197906
Encompass
Reactive Red 120 - Cross-linked 4% agarose beads (Separopore 4B-CL). Specifications: Matrix: Cross-linked 4% agarose beads Dye: Reactive Red-120 Extent of labeling: 3-5 um per mL Supplied: as 50% slurry (0.5M NaCl/0.02% thimerosal) S...
Catalog No. 50-197-906 Supplier Bioworld Supplier No. 201810952
Reactive Red 120 - Cross-linked 4% agarose beads (Separopore 4B-CL). Specifications: Matrix: Cross-linked 4% agarose beads Dye: Reactive Red-120 Extent of labeling: 3-5 um per mL Supplied: as 50% slurry (0.5M NaCl/0.02% thimerosal) S...