Learn More
Bioworld RED SEPAROPORE 4B-CL
Supplier:  Bioworld 201810951
Encompass
This item has a minimum order quantity of 3 per supplier requirements. Reactive Red 120 - Cross-linked 4% agarose beads (Separopore 4B-CL). Specifications: Matrix: Cross-linked 4% agarose beads Dye: Reactive Red-120 Extent of labeling: 3-5 um per mL Supplied: as 50% slurry (0.5M NaCl/0.02% thimerosal) S...
Catalog No. 50-197-905