Learn More
Chemglass Life Sciences Septum Stopper, Sleeve Type, for 5mm OD NMR Tubes, Red
SDP
Catalog No. CG302209
Encompass_Preferred
Septum Stopper, Sleeve Type, for 5mm OD NMR Tubes, Red. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.
Catalog No. CG302209 Supplier Chemglass Life Sciences Supplier No. CG302209
Septum Stopper, Sleeve Type, for 5mm OD NMR Tubes, Red. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.