Learn More
Thermco Products Inc NIST compliant, factory certified glass Spirit thermometer, -20 deg./110 deg.C, 0.5C div, 76mm imm, certified at -10 deg.C, 0 deg.C, 4 deg.C, 25 deg.C, 37 deg.C, 56 deg.C, 105 deg.C, 305mm
Catalog No. NC0321687
Encompass_Preferred
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In- Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case.
Catalog No. NC0321687 Supplier Thermco Products Inc Supplier No. ACC6113SFC
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In- Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case.