Learn More
AdipoGen Red Starch
SDP
Catalog No. 502560088
Encompass_Preferred
Chemical. CAS n.a. Synthetic Red-dyed soluble starch. An excellent soluble substrate for alpha-amylase assay activity. Used for the measurement of enzyme activity, for research, forensic research, biochemical enzyme assays and in vitro diagnostic analysis.
Catalog No. 50-256-0088 Supplier AdipoGen Supplier No. CDXS0035G001
Chemical. CAS n.a. Synthetic Red-dyed soluble starch. An excellent soluble substrate for alpha-amylase assay activity. Used for the measurement of enzyme activity, for research, forensic research, biochemical enzyme assays and in vitro diagnostic analysis.