Learn More
Leica Biosystems Leica Super Cassette, Red - 3 x 2 x ¾ in (75 x 52 x 17 mm), 100 per case
Catalog No. NC3711077
Encompass
Leica Super Cassettes are large-format histology cassettes designed for processing and embedding larger tissue specimens, including whole organs, eyes, intact joints, and other oversized samples. They accommodate larger sections than standard cassettes and are intended for veterinary, research, and pharmaceutical laboratory use. Processing schedules may need to be adjusted for larger specimens.
  • Accommodates large specimens such as whole organs, eyes, and intact joints.
  • Dimensions: 3 x 2 x ¾ in (75 x 52 x 17 mm).
  • Red color for easy rack and specimen identification.
  • Loose-format cassettes supplied 100 per case for convenient inventory handling.
  • Intended for processing and embedding of larger tissue sections in research and veterinary labs.
  • Processing schedules must be adjusted when using larger specimens.
Catalog No. NC3711077 Supplier Leica Biosystems Supplier No. 38VSP59066
Leica Super Cassettes are large-format histology cassettes designed for processing and embedding larger tissue specimens, including whole organs, eyes, intact joints, and other oversized samples. They accommodate larger sections than standard cassettes and are intended for veterinary, research, and pharmaceutical laboratory use. Processing schedules may need to be adjusted for larger specimens.
  • Accommodates large specimens such as whole organs, eyes, and intact joints.
  • Dimensions: 3 x 2 x ¾ in (75 x 52 x 17 mm).
  • Red color for easy rack and specimen identification.
  • Loose-format cassettes supplied 100 per case for convenient inventory handling.
  • Intended for processing and embedding of larger tissue sections in research and veterinary labs.
  • Processing schedules must be adjusted when using larger specimens.