Learn More
As One International Inc Red-Taq DNA Polymerase 2x Master Mix, 2.0 mM MgCl2 (final concentration), 2,500Rxn
Supplier:  As One International Inc AO190306
Encompass_Preferred
AS ONE Taq DNA Polymerase with Red Dye Master Mix is a ready-to-use 2.0x reaction mix. Simply add primers, template, and water to successfully carry out primer extensions and other molecular biology applications. AS ONE Taq Polymerase, the NH4+ buffer system, dNTPs, and magnesium chloride are conveniently present in the Taq DNA Polymerase Mix. An inert red dye and a stabilizer are also present to allow direct loading of the final products onto a gel for analysis. AS ONE Taq DNA Polymerase with Red Dye Mix offers several advantages. Set up time is significantly reduced. There is no need to buy and use separate loading dyes to load reaction products onto agarose gels for electrophoresis and subsequent visualization. The chance of contaminating component stocks is eliminated. Reduction of reagent handling steps leads to better reproducibility. Standard tests can be set up with the confidence that results will be consistent every time.
Catalog No. 50-165-5742