Learn More
Montana Molecular Tau-Targeted Montana Paintbrush Red (Red TauPaint) Expression Kit - Big Sky BacMam, 5mL - Montana Molecular
SDP
Supplier:  Montana Molecular P1050R005
Encompass_Preferred
Bright, red fluorescent protein fused to Tau for live-cell imaging—no fixation, staining, or cell lysis required. Visualize cytoskeletal structures and cell morphology, over hours to days, in living cells. Delivered using Montana Molecular’s Big Sky BacMam vector for enhanced expression across a wide range of cell types, including primary cultures and iPSCs. Ideal for non-destructive studies of Tau organization, cellular phenotypes, and high-content imaging.
Catalog No. NC3787551