Learn More
Andwin Scientific RED TOUGH TAGS LARGE 1000/R
SDP
Supplier:  Andwin Scientific 91871004
Encompass_Preferred
The large red Tough-Tags labels are conveniently pre-cut to be easily put on microcentrifuge tubes and other laboratory containers. The labels can resist temperatures as low as -20C (-80C when used on virgin polypropylene tubes.) This product can also withstand organic solvents and caustic agents without peeling or falling off. The labels measure 1.28125 in W x 0.28125 in H.
Catalog No. 50-821-007