Learn More
Bellco Glass, Inc. Red Tube Marker
SDP
Catalog No. 502330011
Encompass_Preferred
DescriptionFast drying, smudge resistant, fine point markerIdeal for permanently marking all plastic lab ware, especially centrifuge tubes; even works with frozen tubes! Pack Description5/BagCase Quantity5Primary ColorRedAlcohol/Solvent ResistantNo
Catalog No. 50-233-0011 Supplier Bellco Glass, Inc. Supplier No. 229408000
DescriptionFast drying, smudge resistant, fine point markerIdeal for permanently marking all plastic lab ware, especially centrifuge tubes; even works with frozen tubes! Pack Description5/BagCase Quantity5Primary ColorRedAlcohol/Solvent ResistantNo